ENA369382284 Enamine

2H,3H-[1,4]dioxino[2,3-c]pyridine-7-carbaldehyde

  • MDL number MFCD09907902
  • Cas Number : 443955-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-102159
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart