ENA369382286 Enamine

2,2,4-trimethylpentan-1-amine hydrochloride

  • MDL number MFCD22196459
  • Cas Number : 1376325-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102162

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102162
Availability details for
Quantity
  • Availability
  • Add to Cart