ENA369382288 Enamine

3-(1H-imidazol-1-yl)-4-methylpyridine

  • MDL number MFCD22056346
  • Cas Number : 1376156-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-100257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100257
Availability details for
Quantity
  • Availability
  • Add to Cart