ENA369382300 Enamine

[1-(morpholin-4-ylmethyl)cyclohexyl]methanamine

  • MDL number MFCD16700544
  • Cas Number : 1248125-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102180

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102180
Availability details for
Quantity
  • Availability
  • Add to Cart