ENA369382304 Enamine

5-(chloromethyl)-3-(2-phenylethyl)-4,5-dihydro-1,2-oxazole

  • MDL number MFCD19665159
  • Cas Number : 1344286-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102186

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102186
Availability details for
Quantity
  • Availability
  • Add to Cart