ENA369382311 Enamine

2-(3-methoxy-4-nitrophenyl)acetonitrile

  • MDL number MFCD13192129
  • Cas Number : 104103-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102258
Availability details for
Quantity
  • Availability
  • Add to Cart