ENA369382316 Enamine

2-(3,5-dimethylpiperidin-1-yl)cyclohexan-1-one

  • MDL number MFCD12059115
  • Cas Number : 1218135-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102264

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102264
Availability details for
Quantity
  • Availability
  • Add to Cart