ENA369382322 Enamine

4-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)benzoic acid

  • MDL number MFCD17603895
  • Cas Number : 1165931-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241...243
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102275
Availability details for
Quantity
  • Availability
  • Add to Cart