ENA369382327 Enamine

1H,2H,3H,4H-pyrido[3,4-b]pyrazine

  • MDL number MFCD13193470
  • Cas Number : 35808-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102281
Availability details for
Quantity
  • Availability
  • Add to Cart