ENA369382331 Enamine

2-amino-4-(4-methoxyphenyl)-2-methylbutanoic acid

  • MDL number MFCD02662652
  • Cas Number : 1182826-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 303...305
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102285

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102285
Availability details for
Quantity
  • Availability
  • Add to Cart