ENA369382332 Enamine

1-phenyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD16845143
  • Cas Number : 68207-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102286

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102286
Availability details for
Quantity
  • Availability
  • Add to Cart