ENA369382337 Enamine

4-cyanothiophene-2-carboxylic acid

  • MDL number MFCD05664844
  • Cas Number : 406719-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102306

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-102306
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart