ENA369382349 Enamine

N-{3-fluoro-4-[methyl(propan-2-yl)amino]phenyl}prop-2-enamide

  • MDL number MFCD12123355
  • Cas Number : 1156048-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102348
Availability details for
Quantity
  • Availability
  • Add to Cart