ENA369382350 Enamine

2-amino-3-[(2-phenylethyl)sulfanyl]propanoic acid

  • MDL number MFCD09934994
  • Cas Number : 14510-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100275
Availability details for
Quantity
  • Availability
  • Add to Cart