ENA369382351 Enamine

2-(pyridin-2-yl)imidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD14540239
  • Cas Number : 1216060-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102350
Availability details for
Quantity
  • Availability
  • Add to Cart