ENA369382361 Enamine

methyl 2-[3,5-dimethyl-1-(pyridin-2-yl)-1H-pyrazol-4-yl]propanoate

  • MDL number MFCD17929442
  • Cas Number : 1280673-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102362

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102362
Availability details for
Quantity
  • Availability
  • Add to Cart