ENA369382362 Enamine

3-ethylbenzene-1-sulfonyl chloride

  • MDL number MFCD09740557
  • Cas Number : 34586-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102363

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102363
Availability details for
Quantity
  • Availability
  • Add to Cart