ENA369382363 Enamine

N-ethyl-4-sulfanylpyridine-2-carboxamide

  • MDL number MFCD12189520
  • Cas Number : 1154593-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102364

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102364
Availability details for
Quantity
  • Availability
  • Add to Cart