ENA369382375 Enamine

2,6-dibromo-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD11650681
  • Cas Number : 1094671-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102395

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102395
Availability details for
Quantity
  • Availability
  • Add to Cart