ENA369382380 Enamine

1-(3-{[(tert-butoxy)carbonyl]amino}propyl)-1H-indole-5-carboxylic acid

  • MDL number MFCD22196477
  • Cas Number : 1376288-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102402
Availability details for
Quantity
  • Availability
  • Add to Cart