ENA369382381 Enamine

3-amino-1-(3-phenylpropyl)pyrrolidin-2-one

  • MDL number MFCD14689873
  • Cas Number : 1250707-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100304
Availability details for
Quantity
  • Availability
  • Add to Cart