ENA369382386 Enamine

N-(3,3,3-trifluoropropyl)aniline

  • MDL number MFCD12771211
  • Cas Number : 405-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102408
Availability details for
Quantity
  • Availability
  • Add to Cart