ENA369382388 Enamine

5-methylthieno[2,3-d][1,3]thiazol-2-amine

  • MDL number MFCD20706929
  • Cas Number : 41940-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102412

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102412
Availability details for
Quantity
  • Availability
  • Add to Cart