ENA369382392 Enamine

2H,3H,4H-pyrido[4,3-b][1,4]thiazin-3-one

  • MDL number MFCD22196484
  • Cas Number : 18504-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-208
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102417
Availability details for
Quantity
  • Availability
  • Add to Cart