ENA369382395 Enamine

2-(4-chlorophenyl)-2-(1H-pyrazol-1-yl)ethan-1-amine

  • MDL number MFCD16062104
  • Cas Number : 1251104-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102422
Availability details for
Quantity
  • Availability
  • Add to Cart