ENA369382399 Enamine

5-(4-aminophenyl)-1,3,4-oxadiazol-2-ol

  • MDL number MFCD02182673
  • Cas Number : 113118-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102452

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-102452
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart