ENA369382403 Enamine

(3,4-dichloropyridin-2-yl)methanol

  • MDL number MFCD11045050
  • Cas Number : 103949-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-68
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102457
Availability details for
Quantity
  • Availability
  • Add to Cart