ENA369382406 Enamine

N-(1-carbamothioylbutyl)benzamide

  • MDL number MFCD12792747
  • Cas Number : 1283049-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...204
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102460

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102460
Availability details for
Quantity
  • Availability
  • Add to Cart