ENA369382597 Enamine

4-(3-methoxyphenyl)-2-(pyrrolidin-2-yl)-1H-imidazole dihydrochloride

  • MDL number MFCD22196501
  • Cas Number : 1384430-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102609
Availability details for
Quantity
  • Availability
  • Add to Cart