ENA369382612 Enamine

6-oxaspiro[4.5]decan-9-yl methanesulfonate

  • MDL number MFCD22196528
  • Cas Number : 503551-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102869
Availability details for
Quantity
  • Availability
  • Add to Cart