ENA369382614 Enamine

5-fluoro-3-methyl-2-nitrobenzoic acid

  • MDL number MFCD22196532
  • Cas Number : 952479-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102926

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102926
Availability details for
Quantity
  • Availability
  • Add to Cart