ENA369382618 Enamine

2-(aminomethyl)-3-fluoro-N,N-dimethylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD22196539
  • Cas Number : 1384431-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103061
Availability details for
Quantity
  • Availability
  • Add to Cart