ENA369382622 Enamine

4-(3-methyl-1,2,4-oxadiazol-5-yl)cyclohexan-1-amine hydrochloride

  • MDL number MFCD22196544
  • Cas Number : 1384430-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103071

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103071
Availability details for
Quantity
  • Availability
  • Add to Cart