ENA369382623 Enamine

N-[2-(aminomethyl)-3-fluorophenyl]methanesulfonamide hydrochloride

  • MDL number MFCD22196545
  • Cas Number : 1384428-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103080

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103080
Availability details for
Quantity
  • Availability
  • Add to Cart