ENA369382634 Enamine

8-cyclopropyl-3-hydrazinyl-8-azabicyclo[3.2.1]octane dihydrochloride

  • MDL number MFCD22196557
  • Cas Number : 1384430-12-9
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-cyclopropyl-3-hydrazino-8-azabicyclo[3.2.1]octane dihydrochloride
Inchi Code 1S/C10H19N3.2ClH/c11-12-7-5-9-3-4-10(6-7)13(9)8-1-2-8;;/h7-10,12H,1-6,11H2;2*1H
Inchi Key PGVYAVIOELJOKR-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-103160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103160
Availability details for
Quantity
  • Availability
  • Add to Cart