ENA369382637 Enamine

2-aminocyclohexane-1-carboxylic acid hydrochloride

  • MDL number MFCD22196561
  • Cas Number : 1384430-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103185

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103185
Availability details for
Quantity
  • Availability
  • Add to Cart