ENA369382654 Enamine

3,4-dihydrospiro[1-benzopyran-2,1'-cyclobutan]-4-one

  • MDL number MFCD22196586
  • Cas Number : 934554-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103649
Availability details for
Quantity
  • Availability
  • Add to Cart