ENA369382656 Enamine

3-(4,6-dimethoxy-1H-indol-2-yl)propan-1-amine

  • MDL number MFCD22196588
  • Cas Number : 1384428-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103687
Availability details for
Quantity
  • Availability
  • Add to Cart