ENA369382657 Enamine

1H,2H,3H,4H,5H,6H,7H-pyrazolo[3,4-c]pyridin-3-one dihydrochloride

  • MDL number MFCD22196589
  • Cas Number : 1384431-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103737

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103737
Availability details for
Quantity
  • Availability
  • Add to Cart