ENA369382659 Enamine

1-[(1-aminobutan-2-yl)oxy]-2-(trifluoromethyl)benzene hydrochloride

  • MDL number MFCD22196591
  • Cas Number : 1384430-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103740

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103740
Availability details for
Quantity
  • Availability
  • Add to Cart