ENA369382663 Enamine

4-methoxy-5-sulfamoylthiophene-2-carboxylic acid

  • MDL number MFCD22196596
  • Cas Number : 1384429-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254...256
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103798

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103798
Availability details for
Quantity
  • Availability
  • Add to Cart