ENA369382665 Enamine

2-[(6-phenylpyridazin-3-yl)amino]ethan-1-ol

  • MDL number MFCD22196598
  • Cas Number : 80071-91-6
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103859
Availability details for
Quantity
  • Availability
  • Add to Cart