ENA369382673 Enamine

methyl 4-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)benzoate

  • MDL number MFCD22196605
  • Cas Number : 1384430-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104050

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104050
Availability details for
Quantity
  • Availability
  • Add to Cart