ENA369382675 Enamine

ethyl 6-amino-2,3,4-trimethoxybenzoate hydrochloride

  • MDL number MFCD22196607
  • Cas Number : 1384428-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104075

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104075
Availability details for
Quantity
  • Availability
  • Add to Cart