ENA369382678 Enamine

[3-(3,5-dimethoxyphenoxy)propyl](methyl)amine hydrochloride

  • MDL number MFCD22196611
  • Cas Number : 1384430-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120...122
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104160

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104160
Availability details for
Quantity
  • Availability
  • Add to Cart