ENA369382679 Enamine

3-(3,5-dimethoxyphenoxy)propan-1-amine hydrochloride

  • MDL number MFCD22196612
  • Cas Number : 100840-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104161
Availability details for
Quantity
  • Availability
  • Add to Cart