ENA369382737 Enamine

1-(2-methoxyethyl)-5-(trifluoromethyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD22196674
  • Cas Number : 1384427-78-4
  • Molecular Weight : 239.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91004
Availability details for
Quantity
  • Availability
  • Add to Cart