ENA369382749 Enamine

1-(2-chloroethyl)-3,5-dimethyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD22056462
  • Cas Number : 53946-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96010
Availability details for
Quantity
  • Availability
  • Add to Cart