ENA369382750 Enamine

5-(trifluoromethyl)pyridine-2,3-dicarboxylic acid

  • MDL number MFCD22196687
  • Cas Number : 1202634-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96445
Availability details for
Quantity
  • Availability
  • Add to Cart