ENA369382752 Enamine

2-(4H-1,2,4-triazol-3-yl)propan-2-amine dihydrochloride

  • MDL number MFCD22056464
  • Cas Number : 1384427-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96674
Availability details for
Quantity
  • Availability
  • Add to Cart